Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TBX3 Antibody, Alexa Fluor™ 532, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP171853AF532
Description
TBX3 Polyclonal antibody specifically detects TBX3 in Human samples. It is validated for Western BlotSpecifications
TBX3 | |
Polyclonal | |
Western Blot | |
bladder cancer related protein XHL, T-box 3, T-box protein 3, T-box transcription factor TBX3, TBX3-ISO, ulnar mammary syndrome, UMS, XHL | |
The immunogen for this product maps to a region between residue 400 and 450 of human T-box 3 using the numbering given in entry NP_057653.3 (GeneID 6926). | |
0.1 mL | |
Angiogenesis, Apoptosis, Cardiovascular Biology, Cell Biology | |
6926 | |
Store at 4°C in the dark. | |
IgG |
Western Blot | |
Alexa Fluor 532 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction