Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TBX3 Antibody (SR2261), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP32223250UL
Description
TBX3 Monoclonal antibody specifically detects TBX3 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, ImmunoprecipitationSpecifications
TBX3 | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, Flow Cytometry, Immunoprecipitation | |
SR2261 | |
Western Blot 1:500-1:2000, Flow Cytometry 1:100, Immunoprecipitation 1:50 | |
bladder cancer related protein XHL, T-box 3, T-box protein 3, T-box transcription factor TBX3, TBX3-ISO, ulnar mammary syndrome, UMS, XHL | |
A synthesized peptide derived from human TBX3 (Uniprot #: O15119) | |
50 μg | |
Angiogenesis, Apoptosis, Cardiovascular Biology, Cell Biology | |
6926 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction