Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TBX3 Polyclonal Antibody, Bethyl Laboratories

Catalog No. 501564256
Encompass_Preferred
Change view
Click to view available options
20 μL; Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
50-156-4256 20 μL; Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-4256 Supplier Bethyl Laboratories, Inc Supplier No. A303098AT
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt.

T-box 3 (TBX3) is a member of a phylogenetically conserved family of genes that share a common DNA-binding domain, the T-box. T-box genes encode transcription factors involved in the regulation of developmental processes. The Tbx3 protein is a transcriptional repressor that binds the canonical Brachychury bindig sites, and is thought to play a role in the anterior/posterior axis of the tetrapod forelimb. Tbx3 is required for normal mammary development and is also implicated in tumor development. Tbx3 expression also increases during osteoblast differentiation, and may function as a determit of osteoblast cell numbers. Mutations in T-box 3 cause ulnar-mammary syndrome (MIM 181450), affecting limb, apocrine gland, tooth, hair, and genital development. Alternative splicing results in three transcript variants encoding different isoforms; however, the nature of one full length variant has not been determined. Subsets of human breast cancer cell lines overexpress TBX36. in vitro, Tbx3 overexpression in mouse embryo fibroblasts leads to immortalization of cells. Truncated forms of Tbx3 are increased in plasma samples.

Specifications

Antigen TBX3
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene TBX3
Gene Accession No. O15119
Gene Alias T-box transcription factor TBX3, T-box protein 3
Gene Symbols TBX3
Host Species Rabbit
Immunogen Between 400 and 450
Purification Method Antigen affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6926
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.