Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TBX3 Rabbit anti-Mouse, Rat, Clone: 3G9F1, Novus Biologicals™
SDP

Catalog No. p-7248336 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB168785 20 μg
NB168784 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. NB168785 Supplier Novus Biologicals Supplier No. NBP31684820UL
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

TBX3 Monoclonal antibody specifically detects TBX3 in Mouse, Rat samples. It is validated for Western Blot

Specifications

Antigen TBX3
Applications Western Blot
Classification Monoclonal
Clone 3G9F1
Conjugate Unconjugated
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias bladder cancer related protein XHL, T-box 3, T-box protein 3, T-box transcription factor TBX3, TBX3-ISO, ulnar mammary syndrome, UMS, XHL
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 644-743 of human TBX3 (O15119).
Purification Method Affinity purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Angiogenesis, Apoptosis, Cardiovascular Biology, Cell Biology
Primary or Secondary Primary
Gene ID (Entrez) 6926
Target Species Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.