Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TBX3 Recombinant Rabbit Monoclonal Antibody (JE33-38)
GREENER_CHOICE

Catalog No. FEMA550714
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEMA550714 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEMA550714 Supplier Invitrogen™ Supplier No. MA550714
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Positive control: JAR cell lysates, HepG2. Predicted band size: 79 kDa Subcellular Location: Nucleus.

T-box 3 (TBX3) is a member of a phylogenetically conserved family of genes that share a common DNA-binding domain, the T-box. T-box genes encode transcription factors involved in the regulation of developmental processes. The Tbx3 protein is a transcriptional repressor that binds the canonical Brachychury bindig sites, and is thought to play a role in the anterior/posterior axis of the tetrapod forelimb. Tbx3 is required for normal mammary development and is also implicated in tumor development. Tbx3 expression also increases during osteoblast differentiation, and may function as a determinant of osteoblast cell numbers. Mutations in T-box 3 cause ulnar-mammary syndrome (MIM 181450), affecting limb, apocrine gland, tooth, hair, and genital development. Alternative splicing results in three transcript variants encoding different isoforms; however, the nature of one full length variant has not been determined. Subsets of human breast cancer cell lines overexpress TBX36. in vitro, Tbx3 overexpression in mouse embryo fibroblasts leads to immortalization of cells. Truncated forms of Tbx3 are increased in plasma samples.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TBX3
Applications Flow Cytometry, Western Blot
Classification Recombinant Monoclonal
Clone JE33-38
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
Gene Tbx3
Gene Accession No. O15119
Gene Alias bladder cancer related protein XHL; D5Ertd189e; T-box 3; T-box 3 (ulnar mammary syndrome); T-box protein 3; T-box transcription factor TBX3; TBX3; TBX3-ISO; UMS; XHL
Gene Symbols Tbx3
Host Species Rabbit
Immunogen Synthetic peptide within Human Tbx3 aa 694-743/743.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6926
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.