Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TBX4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA565383
Description
Immunogen sequence: KEYPVISKSIM RQRLISPQLS ATPDVGPLLG THQALQHYQH ENGAHSQLAE PQDLPLSTFP TQRDSSLFYH CLKRRADGTR HLDLPCKRSY LEAPSSVGED HYFRSPPPYD QQMLSPS Highest antigen sequence identity to the following orthologs - mouse 85%, rat 86%.
This gene is a member of a phylogenetically conserved family of genes that share a common DNA-binding domain, the T-box. T-box genes encode transcription factors involved in the regulation of developmental processes. This gene is the human homolog of mouse Tbx4, which is closely linked to Tbx2 on mouse chromosome 11. Similarly this gene, like TBX2, maps to human chromosome 17. Expression studies in mouse and chicken show that Tbx4 is expressed in developing hindlimb, but not in forelimb buds, suggesting a role for this gene in regulating limb development and specification of limb identity.
Specifications
TBX4 | |
Polyclonal | |
Unconjugated | |
TBX4 | |
3930401C23; ICPPS; SPS; T-box 4; T-box 4 protein; T-box protein 4; T-box transcription factor 4; T-box transcription factor TBX4; Tbx4; transcription factor Tbx4 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
9496 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P57082 | |
TBX4 | |
Recombinant Human TBX4. Recombinant protein control fragment (Product #RP-106069). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction