Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TBX5 Antibody, PerCP, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299529PCP
Description
TBX5 Polyclonal antibody specifically detects TBX5 in Mouse, Rat samples. It is validated for Immunohistochemistry (Paraffin)Specifications
TBX5 | |
Polyclonal | |
Immunohistochemistry-Paraffin | |
HOS, T-box 5, T-box protein 5, T-box transcription factor TBX5 | |
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human TBX5 | |
0.1 mL | |
Primary | |
Mouse, Rat | |
Purified |
Immunohistochemistry (Paraffin) | |
PerCP | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
6910 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction