Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TBX5 Monoclonal Antibody (7B11)
GREENER_CHOICE

Catalog No. PIMA515526
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA515526 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA515526 Supplier Invitrogen™ Supplier No. MA515526
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA5-15526 targets TBX5 in indirect ELISA, WB applications and shows reactivity with Human samples. The MA5-15526 immunogen is purified recombinant fragment of TBX5 expressed in E. Coli. MA5-15526 detects TBX5 which has a predicted molecular weight of approximately 53kDa.

This gene is a member of a phylogenetically conserved family of genes that share a common DNA-binding domain, the T-box. T-box genes encode transcription factors involved in the regulation of developmental processes. This gene is closely linked to related family member T-box 3 (ulnar mammary syndrome) on human chromosome 12. The encoded protein may play a role in heart development and specification of limb identity. Mutations in this gene have been associated with Holt-Oram syndrome, a developmental disorder affecting the heart and upper limbs. Several transcript variants encoding different isoforms have been described for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TBX5
Applications ELISA, Western Blot
Classification Monoclonal
Clone 7B11
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation Ascites with 0.03% sodium azide
Gene TBX5
Gene Accession No. Q99593
Gene Alias HOS; T-box 5; T-box protein 5; T-box transcription factor TBX5; TBX5
Gene Symbols TBX5
Host Species Mouse
Immunogen Purified recombinant fragment of TBX5 expressed in E. Coli.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6910
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.