Learn More
Invitrogen™ TBX6 Polyclonal Antibody

Description
Store as concentrated solution. Centrifuge briefly prior to opening vial.
Specifications
Specifications
| Antigen | TBX6 |
| Applications | Immunohistochemistry (PFA fixed) |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 20% glycerol and 0.01% thimerosal; pH 7 |
| Gene | TBX6 |
| Gene Accession No. | Q8JIS6 |
| Gene Alias | rib-vertebrae; rv; SCDO5; T-box 6; T-box protein 6; T-box transcription factor 6; T-box transcription factor TBX6; TBX6 |
| Gene Symbols | TBX6 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.