Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TBXA2R Polyclonal Antibody, Invitrogen™

Catalog No. PIPA528753
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA528753 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA528753 Supplier Thermo Scientific Supplier No. PA528753

Rabbit Polyclonal Antibody

Recommended positive controls: HeLaS3, HepG2, Raji. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

The thromboxane A2 receptor (TBXA2R) is a member of the Prostanoid Receptor subfamily. TBXA2R performs an essential role in hemostasis by interacting with thromboxane A2 to induce platelet aggregation. Thromboxane A2 has been implicated in myocardial infarct, stroke, and asthma. Additionally, thromboxane-prostanoid receptors have been implicated in the pathogenesis of cardiovascular diseases. Changes in TBXA2R function have also been linked to a hereditary bleeding disorder.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TBXA2R
Applications Western Blot
Classification Polyclonal
Concentration 0.55 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Gene TBXA2R
Gene Accession No. P21731
Gene Alias BDPLT13; prostanoid TP receptor; TBXA2R; Thromboxane A2 receptor; thromboxane prostanoid; Thromboxane receptor; TP; TXA2 receptor; TXA2-R; TXR2
Gene Symbols TBXA2R
Host Species Rabbit
Immunogen Synthetic peptide corresponding to a region within amino acids 1 and 45 of Human TXA2R
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6915
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.