Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ TC 2559 difumarate
GREENER_CHOICE

Catalog No. 273710
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
27-371-0 10 mg
27-375-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 27-371-0 Supplier Tocris Bioscience™ Supplier No. 2737/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Selective partial agonist at α4β2 receptors

Subtype-selective partial agonist for α4β2 nicotinic acetylcholine receptors (EC50 values are 0.18, 12.5, 14.0, > 30, > 100 and > 100 μM for α4β2, α4β4, α2β4, α3β4, α3β2 and α7 receptor subtypes respectively). Displays selectivity for (α4)2(β2)3 receptor stoichiometry and enhanced CNS-PNS selectivity ratio. Attenuates scopolamine-induced cognitive deficits in a step-through passive avoidance task.

Chemical Identifiers

CAS 212332-35-9
Molecular Formula C20H26N2O9

Specifications

Quantity 10 mg
Formula Weight 438.43
Purity >98%
Molecular Weight (g/mol) 438.433
Chemical Name or Material TC 2559 difumarate
Recommended Storage Desiccate at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.