Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ TC-A 2317 hydrochloride
GREENER_CHOICE

Catalog No. 406610
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
40-661-0 10 mg
40-665-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 40-661-0 Supplier Tocris Bioscience™ Supplier No. 4066/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent, selective Aurora kinase A inhibitor

Potent Aurora kinase A inhibitor (Ki = 1.2 nM compared to 101 nM for inhibition of Aurora kinase B). Selective over 60 other kinases (IC50 values > 1000 nM). Exhibits good cell permeability and antitumor activity.

Chemical Identifiers

CAS 1245907-03-2
Molecular Formula C19H29ClN6O

Specifications

Quantity 10 mg
Formula Weight 392.93
Purity >98%
Molecular Weight (g/mol) 392.932
Chemical Name or Material TC-A 2317 hydrochloride
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.