Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ TC-DAPK 6
GREENER_CHOICE

Catalog No. 430110
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
43-011-0 10 mg
43-015-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 43-011-0 Supplier Tocris Bioscience™ Supplier No. 4301/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent and selective inhibitor of DAPK1

Potent and selective, ATP-competitive inhibitor of death-associated protein kinase 1 (DAPK1) (IC50 values are 69 and 225 nM for DAPK1 and DAPK3 respectively, when assayed with 10 μM ATP). Displays selectivity for DAPK1 over a range of 48 other kinases, including Abl, AMPK, Chk1, Met and Src (IC50 > 10 μM).

Chemical Identifiers

CAS 315694-89-4
Molecular Formula C17H12N2O2

Specifications

Quantity 10 mg
Formula Weight 276.29
Purity >98%
Molecular Weight (g/mol) 276.295
Chemical Name or Material TC-DAPK 6
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.