Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ TC-E 5002
GREENER_CHOICESDP

Catalog No. p-5546060 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-895-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 50-895-0 Supplier Tocris Bioscience™ Supplier No. 5089/50
Add to Cart
Add to Cart
This item is not returnable. View return policy

Selective KDM2/7 inhibitor

TC-E 5002 is a selective histone demethylase KDM2/7 subfamily inhibitor (IC50 values are 0.2, 1.2, 6.8, 55, 83, >100 and >120 μM for KDM7A, KDM7B, KDM2A, KDM5A, KDM4C, KDM6A and KDM4A respectively). Inhibits growth of HeLa and KYSE-150 cancer cells in vitro.

Specifications

Quantity 50 mg
Product Type TC-E 5002
Purity >98%
Molecular Weight (g/mol) 285.38
Inhibitors TC-E 5002

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.