Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ TC-F 2
GREENER_CHOICESDP

Catalog No. 435550 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
43-555-0 50 mg
43-551-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 43-555-0 Supplier R&D Systems™ Supplier No. 4355/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Potent, reversible and selective FAAH inhibitor

Potent, reversible inhibitor of fatty acid amide hydrolase (FAAH) (IC50 values are 28 and 100 nM for human and rat FAAH respectively). Noncovalent inhibitor; displays selectivity for FAAH over cannabinoid-related targets (IC50 > 20 μM for CB1, CB2 and TRPV1). Active in vivo.

Chemical Identifiers

CAS 1304778-15-1
Molecular Formula C26H25N5O2

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 439.51
Purity >99%
Molecular Weight (g/mol) 439.519
Chemical Name or Material TC-F 2
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.