Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ TC-G 1001
GREENER_CHOICE

Catalog No. 487910
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
48-791-0 10 mg
48-795-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 48-791-0 Supplier Tocris Bioscience™ Supplier No. 4879/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

GPR35 agonist; more potent than zaprinast (Cat. No. 0947)

GPR35 agonist (pEC50 values are 7.59 and 8.36 for β-arrestin and Gαq-i5 Ca2+ assays respectively). Displays 1000-fold greater potency at human GPR35 receptors relative to mouse and rat GPR35 orthologs in an IP1 accumulation assay. More potent than zaprinast (Cat. No. 0947).

Chemical Identifiers

CAS 494191-73-0
Molecular Formula C17H11FN2O3S

Specifications

Quantity 10 mg
Formula Weight 342.34
Purity >98%
Molecular Weight (g/mol) 342.344
Chemical Name or Material TC-G 1001
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.