Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Description
GPR35 agonist (pEC50 values are 7.59 and 8.36 for β-arrestin and Gαq-i5 Ca2+ assays respectively). Displays 1000-fold greater potency at human GPR35 receptors relative to mouse and rat GPR35 orthologs in an IP1 accumulation assay. More potent than zaprinast (Cat. No. 0947).
Specifications
Specifications
| Quantity | 50 mg |
| Formula Weight | 342.34 |
| Purity | >98% |
| Molecular Weight (g/mol) | 342.344 |
| Chemical Name or Material | TC-G 1001 |
Safety and Handling
| Recommended Storage | Store at +4°C |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?

