Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ TC-MCH 7c
GREENER_CHOICESDP

Catalog No. 436510 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
43-651-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 43-651-0 Supplier R&D Systems™ Supplier No. 4365/10

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Selective melanin-concentrating hormone receptor 1 (MCH1R) antagonist

Potent, selective melanin-concentrating hormone receptor 1 (MCH1R) antagonist (IC50= 5.6 nM in hMCH1R-expressing CHO cells). Displays selectivity for MCH1R over MCH2R (IC50 = > 10 μM). Decreases body weight in a mouse model of diet-induced obesity. Brain penetrant; orally active.

Chemical Identifiers

CAS 864756-35-4
Molecular Formula C24H25FN2O3

Specifications

Quantity 10 mg
Product Line Tocris
Formula Weight 408.47
Purity >97%
Molecular Weight (g/mol) 408.473
Chemical Name or Material TC-MCH 7c
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.