Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ TC NTR1 17
GREENER_CHOICESDP

Catalog No. 508750RD Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-875-0RD 50 mg
50-871-0RD 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 50-875-0RD Supplier Tocris Bioscience™ Supplier No. 5087/50
Add to Cart
Add to Cart
This item is not returnable. View return policy

Selective non-peptide NTS1 agonist

TC NTR1 17 is a non-peptide neurotensin receptor 1 (NTS1) partial agonist (EC50 <15.6 nM and Emax = 63% of neurotensin response in Ca2+ mobilization assay). Exhibits over 50-fold selectivity for NTS1 over NTS2 and GPR35.

Specifications

Quantity 50 mg
Product Type TC NTR1 17
Purity >99%
Molecular Weight (g/mol) 522.98

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.