Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ TC OT 39
GREENER_CHOICESDP

Catalog No. 462510 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
46-251-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 46-251-0 Supplier R&D Systems™ Supplier No. 4625/10

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Potent non-peptide oxytocin receptor partial agonist; also V1a receptor antagonist

TC OT 39 is a potent non-peptide oxytocin receptor partial agonist (EC50 values are 33 nM and 850 nM for the oxytocin receptor and V2 vasopressin receptor respectively). Also a V1a vasopressin receptor antagonist (Ki = 330 nM). Active in vivo in a rat model of uterine response.

Specifications

Quantity 10 mg
Product Line Tocris
Product Type TC OT 39
Purity >97%
Molecular Weight (g/mol) 600.78
Inhibitors TC OT 39

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.