Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TC-PTP/PTPN2 Antibody (001), DyLight 350, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290722UV
Description
TC-PTP/PTPN2 Monoclonal specifically detects TC-PTP/PTPN2 in Mouse samples. It is validated for ELISA.Specifications
TC-PTP/PTPN2 | |
Monoclonal | |
DyLight 350 | |
protein tyrosine phosphatase, non-receptor type 2, PTPTPTN2, T-cell protein tyrosine phosphatase, T-cell protein-tyrosine phosphatase, TCELLPTP, TC-PTP, TCPTPEC 3.1.3.48, tyrosine-protein phosphatase non-receptor type 2 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse TC-PTP/PTPN2 (Q06180-1; Ser2-Asn314). | |
0.1 mL | |
Primary | |
Mouse | |
Purified |
ELISA | |
1 | |
50 mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
5771 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction