Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ TC-S 7010

Catalog No. 528650
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
52-865-0 50 mg
52-861-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 52-865-0 Supplier Tocris Bioscience™ Supplier No. 5286/50
This item is not returnable. View return policy

Potent and selective Aurora kinase A inhibitor

TC-S 7010 is a potent and selective inhibitor of Aurora kinase A (IC50 = 3.4 nM); exhibits 1000-fold selectivity for Aurora kinase A over Aurora kinase B. Displays antiproliferative activity in HCT116 and HT29 cells (IC50 values are 0.19 and 2.9 μM respectively).

Specifications

Chemical Name or Material N-(2-Chlorophenyl)-4-[[2-[[4-[2-(4-ethyl-1-piperazinyl)-2-oxoethyl]phenyl]amino]-5-fluoro-4-pyrimidinyl]amino]-benzamide
CAS 1158838-45-9
Quantity 50 mg
Target Aurora Kinase Inhibitors
Molecular Formula C31H31ClFN7O2
Formula Weight 588.07
Purity >99%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.