Learn More
Invitrogen™ TC10 Polyclonal Antibody

Description
This antibody is predicted to react with bovine, chicken, canine, equine, mouse, porcine, non-human primate, rat and yeast based on sequence homology.
Specifications
Specifications
| Antigen | TC10 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | RHOQ |
| Gene Accession No. | P17081 |
| Gene Alias | ARHQ; epididymis secretory protein Li 42; HEL-S-42; ras homolog family member Q; ras homolog gene family, member Q; RASL7A; ras-like protein family member 7A; Ras-like protein TC10; RAS-like, family 7, member A; ras-related GTP-binding protein TC10; RHOQ; Rho-related GTP-binding protein RhoQ; small GTPase Tc10; Tc10; TC10A |
| Gene Symbols | RHOQ |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.