Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TC10 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5111209
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5111209 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5111209 Supplier Invitrogen™ Supplier No. PA5111209
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: FDEAIIAILT PKKHTVKKRI GSRCINCCLI T Highest antigen sequence indentity to the following orthologs: Rat - 100%, Mouse - 100%.

TC10 is a member of the RAS superfamily of small GTP-binding proteins (see HRAS, MIM 190020) involved in insulin-stimulated glucose uptake.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TC10
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RHOQ
Gene Accession No. P17081
Gene Alias ARHQ; epididymis secretory protein Li 42; HEL-S-42; ras homolog family member Q; ras homolog gene family, member Q; RASL7A; ras-like protein family member 7A; Ras-like protein TC10; RAS-like, family 7, member A; ras-related GTP-binding protein TC10; RHOQ; Rho-related GTP-binding protein RhoQ; small GTPase Tc10; Tc10; TC10A
Gene Symbols RHOQ
Host Species Rabbit
Immunogen Recombinant Protein corresonding to Human TC10. Recombinant protein control fragment (Product #RP-106080).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23433
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.