Learn More
Mettler Toledo™ TC100 Combicooler Cooling System for use with DSC 5+
Catalog No. 01338207 Shop All METTLER TOLEDO Products
Change view
Click to view available options
For Use With (Equipment):
DSC 5+
Catalog No. For Use With (Equipment)
01-338-207 DSC 5+
1 options
Catalog No. 01-338-207 Supplier Mettler Toledo™ Supplier No. 30857474
Add to Cart
Add to Cart
Request a Quote
Huber cooling system for use with the DSC 5+. Appropriate for use in conjunction with the 700C DSC5+ equipped with combicooler functionality.
Temperature range of -100 to 40°C.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Cooling System
For Use With (Equipment) DSC 5+
Temperature Range (English) -100°C to 40°C