Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TC21/R-Ras2 Antibody, Alexa Fluor™ 405, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP281761AF405
This item is not returnable.
View return policy
Description
TC21/R-Ras2 Polyclonal antibody specifically detects TC21/R-Ras2 in Human, Mouse samples. It is validated for Western Blot, ELISASpecifications
TC21/R-Ras2 | |
Polyclonal | |
50mM Sodium Borate | |
Rabbit | |
Peptide affinity purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Western Blot, ELISA | |
Alexa Fluor 405 | |
ras-related protein R-Ras2, related RAS viral (r-ras) oncogene homolog 2, TC21Ras-like protein TC21, Teratocarcinoma oncogene | |
Rabbit polyclonal R-Ras2 antibody was raised against a 16 amino acid peptide near the carboxy terminus of human R-Ras2. The immunogen is located within the last 50 amino acids of R-Ras2. Amino Acid Squence: PPSPEPTRKEKDKKG | |
0.1 mL | |
Signal Transduction | |
22800 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction