Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TC21/R-Ras2 Antibody (EM-50), DyLight 755, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP262216IR
Description
TC21/R-Ras2 Monoclonal specifically detects TC21/R-Ras2 in Human, Monkey samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence, Flow (Intracellular).Specifications
TC21/R-Ras2 | |
Monoclonal | |
DyLight 755 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
RRAS2 | |
Human TC21 | |
0.1 mL | |
Primary | |
The antibody MEM-31 recognizes a conformationally-dependent epitope of CD8, a cell surface glycoprotein found on most cytotoxic T lymphocytes that mediates efficient cell-cell interactions within the immune system. CD8 is a disulfide-linked dimer and exists as a CD8 alpha/alpha homodimer or CD8 alpha/beta heterodimer (each monomer approx. 32-34 kDa). The antibody does not react with formaldehyde-fixed cells; negative in Western Blotting application. | |
Store at 4°C in the dark. |
Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence | |
EM-50 | |
Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence, Flow (Intracellular) | |
ras-related protein R-Ras2, related RAS viral (r-ras) oncogene homolog 2, TC21Ras-like protein TC21, Teratocarcinoma oncogene | |
Mouse | |
Protein A purified | |
Signal Transduction | |
22800 | |
Human, Monkey | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction