Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TC2N Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA555651
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA555651 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA555651 Supplier Invitrogen™ Supplier No. PA555651
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: IKSCCGGCFY GETEKHNFSV ERDFKAAVPN SQNATISVPP LTSVSVKPQL GCTEDYLLSK LPSDGKEVPF VVPKFKLSYI QPRTQETPSH LEELEGSARA SFGDRKVELS SSSQHGPSYD VYNPFYMYQH ISPDLSRRF Highest antigen sequence identity to the following orthologs: Mouse - 80%, Rat - 78%.

The TC2N gene encodes the Tandem C2 domains nuclear protein, which is characterized by the presence of two C2 domains, C2A and C2B, at the C-terminus. These domains are structured as an eight-stranded antiparallel beta-sandwich, initially recognized as a Ca2+ binding module. TC2N's role varies significantly across different types of cancer. It has been identified as an oncogene in lung, gastric, and hepatocellular carcinomas, where its overexpression promotes cell proliferation, migration, and metastasis. Conversely, in breast cancer, TC2N functions as a tumor suppressor, inhibiting distant metastasis and stemness via the PI3K-Akt signaling pathway. These dual roles highlight its complex involvement in tumorigenesis, with its impact being highly context-dependent on the cancer type.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TC2N
Applications Western Blot, Immunocytochemistry, Western Blot
Classification Polyclonal
Concentration 0.10 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TC2N
Gene Accession No. Q8N9U0
Gene Alias 4933406D09Rik; C14orf47; C2 calcium-dependent domain containing 1; C2CD1; membrane targeting (tandem) C2 domain containing 1; membrane targeting tandem C2 domain-containing protein 1; MTAC2D1; Tac2-N; Tandem C2 domains nuclear protein; tandem C2 domains, nuclear; tandem C2 protein in nucleus; TC2N
Gene Symbols TC2N
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TC2N. Recombinant protein control fragment (Product #RP-88889).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 123036
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.