Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TCAP Antibody (OTI11H10), Alexa Fluor™ 350, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274471AF350
Description
TCAP Monoclonal specifically detects TCAP in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry.Specifications
TCAP | |
Monoclonal | |
Alexa Fluor 350 | |
50 mM sodium borate with 0.05% sodium azide | |
TCAP | |
Full length recombinant protein of human TCAP (NP_003664) produced in E.coli. | |
0.1 mL | |
8557.0 | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry | |
OTI11H10 | |
Western Blot, Immunohistochemistry | |
19 kDa sarcomeric protein, CMD1NTitin cap protein, LGMD2G, limb girdle muscular dystrophy 2G (autosomal recessive), T-cap, TELE, telethonin, titin-cap (telethonin) | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction