Learn More
Invitrogen™ TCAP Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA593208
Description
Immunogen sequence: MATSELSCEV SEENCERREA FWAEWKDLTL STRPEEGCSL HEEDTQRHET YHQQGQCQVL VQRSPWLMMR MGILGRGLQE YQLPYQRVLP LPIFTPAKMG ATKEEREDTP IQLQELLALE TALGGQCVDR QEVAEITKQL PPVVPVSKPG ALRRSLSRSM SQEAQRG; Positive Samples: Mouse heart, Mouse skeletal muscle, Rat heart, Rat skeletal muscle, Rat skeletal muscle; Cellular Location: Cytoplasm, myofibril, sarcomere.
Sarcomere assembly is regulated by the muscle protein titin. Titin is a giant elastic protein with kinase activity that extends half the length of a sarcomere. It serves as a scaffold to which myofibrils and other muscle related proteins are attached. TCAP is a protein found in striated and cardiac muscle that binds to the titin Z1-Z2 domains and is a substrate of titin kinase, interactions thought to be critical to sarcomere assembly. Mutations in TCAP gene are associated with limb-girdle muscular dystrophy type 2G.Sarcomere assembly is regulated by the muscle protein titin. Titin is a giant elastic protein with kinase activity that extends half the length of a sarcomere. It serves as a scaffold to which myofibrils and other muscle related proteins are attached. This gene encodes a protein found in striated and cardiac muscle that binds to the titin Z1-Z2 domains and is a substrate of titin kinase, interactions thought to be critical to sarcomere assembly. Mutations in this gene are associated with limb-girdle muscular dystrophy type 2G.
Specifications
| TCAP | |
| Polyclonal | |
| Unconjugated | |
| Tcap | |
| 19 kDa sarcomeric protein; AI503981; AJ223855; CMD1N; CMH25; LGMD2G; limb girdle muscular dystrophy 2G (autosomal recessive); Tcap; T-cap; TELE; telethonin; teneurin C-terminal associated peptide; titin cap protein; titin-cap; titin-cap (telethonin) | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 8557 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 2.43 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O15273 | |
| Tcap | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-167 of human TCAP (NP_0036641). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.