Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ DCAF12 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5109619
Description
Immunogen sequence: MARKVVSRKR KAPASPGAGS DAQGPQFGWD HSLHKRKRLP PVKRSLVYYL KNREVRLQNE TSYSRVLHGY AAQQLPSLLK EREFH.
Cancer-testis (CT) antigens are a type of antigen whose expression is highly restricted, primarily in normal testis, but aberrantly in different tumor types. Their immunogenicity in cancer patients makes them prime targets for immunotherapy of human tumors. One such CT antigen is TCC52, a novel centrosomal protein. TCC52 is a WD repeat-containing protein that interacts with the COP9 signalosome, a macromolecular complex that interacts with cullin-RING E3 ligases and regulates their activity by hydrolyzing cullin-Nedd8 conjugates. The normal biological function of this protein is not known at this time.
Specifications
| DCAF12 | |
| Polyclonal | |
| Unconjugated | |
| DCAF12 | |
| 1500001L20Rik; 5830424K06Rik; AA420338; AI851081; cancer/testis antigen 102; centrosome-related protein TCC52; CT102; Dcaf12; DDB1 and CUL4 associated factor 12; DDB1- and CUL4-associated factor 12; Kiaa1892; LOC682812; similar to WD repeat domain 40A; TCC52; Testis cancer centrosome-related protein; WD repeat domain 40A; WD repeat-containing protein 40A; WDR40A | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 25853, 682812 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.75 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q5T6F0 | |
| DCAF12 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-85 of human DCAF12 (NP_056212.1). | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction