Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DCAF12 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA520939
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA520939 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA520939 Supplier Invitrogen™ Supplier No. PA520939
Add to Cart
Add to Cart

Chicken Polyclonal Antibody

A suggested positive control is 293 cell lysate. PA5-20939 can be used with blocking peptide PEP-1053.

Cancer-testis (CT) antigens are a type of antigen whose expression is highly restricted, primarily in normal testis, but aberrantly in different tumor types. Their immunogenicity in cancer patients makes them prime targets for immunotherapy of human tumors. One such CT antigen is TCC52, a novel centrosomal protein. TCC52 is a WD repeat-containing protein that interacts with the COP9 signalosome, a macromolecular complex that interacts with cullin-RING E3 ligases and regulates their activity by hydrolyzing cullin-Nedd8 conjugates. The normal biological function of this protein is not known at this time.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DCAF12
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene DCAF12
Gene Accession No. Q5T6F0, Q8BGZ3
Gene Alias 1500001L20Rik; 5830424K06Rik; AA420338; AI851081; cancer/testis antigen 102; centrosome-related protein TCC52; CT102; Dcaf12; DDB1 and CUL4 associated factor 12; DDB1- and CUL4-associated factor 12; Kiaa1892; LOC682812; similar to WD repeat domain 40A; TCC52; Testis cancer centrosome-related protein; WD repeat domain 40A; WD repeat-containing protein 40A; WDR40A
Gene Symbols DCAF12
Host Species Chicken
Immunogen A 15 amino acid peptide near the carboxy terminus of human TCC52.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25853, 682812, 68970
Target Species Human, Mouse, Rat
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgY
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.