Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TCEA1 Rabbit anti-Human, Mouse, Rat, Clone: S02-6C9, Novus Biologicals™

Rabbit Monoclonal Antibody
$263.00 - $573.00
Specifications
Antigen | TCEA1 |
---|---|
Clone | S02-6C9 |
Applications | Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
TCEA1 Monoclonal antibody specifically detects TCEA1 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
TCEA1 | |
Western Blot | |
Unconjugated | |
Rabbit | |
DNA replication Transcription Translation and Splicing | |
50nM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
6917 | |
IgG | |
Affinity purified |
S02-6C9 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
GTF2Stranscription elongation factor A protein 1, SII, TF2STCEA, TFIISTranscription elongation factor TFIIS.o, transcription elongation factor A (SII), 1, Transcription elongation factor S-II protein 1 | |
Recombinant protein of human TCEA1 (Uniprot#: P23193) | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title