Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TCEA1 Recombinant Rabbit Monoclonal Antibody (JE64-90)
GREENER_CHOICE

Catalog No. PIMA544722
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA544722 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA544722 Supplier Invitrogen™ Supplier No. MA544722
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

TCEA1 Recombinant Monoclonal Antibody for Western Blot, IHC (P)

TCEA1 is necessary for efficient RNA polymerase II transcription elongation past template-encoded arresting sites. TCEA1 has a role in suppression of transient pausing, which is the most important contribution of TCEA1 to elongation from a stall position. TCEA1 fuse to PLAG1 protein which is overexpressed in epithelial, myoepithelial, and mesenchymal-like tumor cells in tumors.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TCEA1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Recombinant Monoclonal
Clone JE64-90
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
Gene TCEA1
Gene Accession No. P10711, P23193, Q4KLL0
Gene Alias GTF2S; SII; S-II; TCEA; Tcea1; Tceat; TF2S; TFIIS; transcription elongation factor A (SII) 1; transcription elongation factor A (SII), 1; transcription elongation factor A protein 1; Transcription elongation factor A protein 1-like protein; transcription elongation factor A1; transcription elongation factor S-II protein 1; Transcription elongation factor TFIIS.o; transcription factor IIS
Gene Symbols TCEA1
Host Species Rabbit
Immunogen Synthetic peptide within human TCEA1 aa 201-250/301.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 21399, 362479, 6917
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.