Learn More
Invitrogen™ TCEA2 Polyclonal Antibody

Description
Immunogen sequence: MMGKEEEIAR IARRLDKMVT KKSAEGAMDL LRELKAMPIT LHLLQSTRVG Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 92%.
Specifications
Specifications
| Antigen | TCEA2 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.05 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | TCEA2 |
| Gene Accession No. | Q15560 |
| Gene Alias | AI326274; Protein S-II-T1; Siit1; S-II-T1; SII-T1; Tcea2; Tceat; testis-specific S-II; TFIIS; transcription elongation factor A (SII), 2; transcription elongation factor A protein 2; transcription elongation factor A2; transcription elongation factor S-II protein 2; transcription elongation factor TFIIS.1; transcription elongation factor TFIIS.l; zgc:56325 |
| Gene Symbols | TCEA2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.