Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TCEA2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA540716
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA540716 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA540716 Supplier Invitrogen™ Supplier No. PA540716
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: QSTRVGMSVN ALRKQSSDEE VIALAKSLIK SWKKLLDASD AKARERGRGM Sequence homology: Cow: 86%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 93%; Rabbit: 79%; Rat: 93%; Zebrafish: 86%.

The protein encoded by TCEA2 is found in the nucleus, where it functions as an SII class transcription elongation factor. Elongation factors in this class are responsible for releasing RNA polymerase II ternary complexes from transcriptional arrest at template-encoded arresting sites. The encoded protein has been shown to interact with general transcription factor IIB, a basal transcription factor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TCEA2
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene TCEA2
Gene Accession No. Q15560
Gene Alias AI326274; Protein S-II-T1; Siit1; S-II-T1; SII-T1; Tcea2; Tceat; testis-specific S-II; TFIIS; transcription elongation factor A (SII), 2; transcription elongation factor A protein 2; transcription elongation factor A2; transcription elongation factor S-II protein 2; transcription elongation factor TFIIS.1; transcription elongation factor TFIIS.l; zgc:56325
Gene Symbols TCEA2
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human TCEA2 (aa 45-94).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6919
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.