Learn More
Invitrogen™ TCEA3 Monoclonal Antibody (2C9)

Description
Peptide Sequence: MGQEEELLRI AKKLEKMVAR KNTEGALDLL KKLHSCQMSI QLLQTTRIGV AVNGVRKHCS DKEVVSLAKV LIKNWKRLLD SPGPPKGEKG EEREKAKKKE KGLECSDWKP EAGLSPPRKK REDPKTRRDS VDSKSSASSS PKRPSVERSN SSKSKAESPK TPSSPLTPTF ASSMCLLAPC YLTGDSVRDK CVEMLSAALK ADDDYKDYGV NCDKMASEIE DHIYQELKST DMKYRNRVRS RISNLKDPRN PGLRRNVLSG AISAGLIAKM TAEEMASDEL RELRNAMTQE AIREHQMAKT GGTTTDLFQC SKCKKKNCTY NQVQTRSADE PMTTFVLCNE CGNRWKFC.
Specifications
Specifications
| Antigen | TCEA3 |
| Applications | ELISA, Immunoprecipitation, Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | 2C9 |
| Concentration | 0.45 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | TCEA3 |
| Gene Accession No. | O75764 |
| Gene Alias | hypothetical protein LOC402983; MGC137536 protein; rhabdomyosarcoma antigen MU-RMS-40.22; S-II; SII-K1; Tcea3; TFIIS; TFIIS.H; Tfiish; tissue-specific transcription factor S-II; transcription elongation factor A (SII), 3; transcription elongation factor A protein 3; transcription elongation factor A3; transcription elongation factor S-II protein 3; Transcription elongation factor TFIIS.h; zgc:77732 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.