Learn More
Invitrogen™ TCEA3 Polyclonal Antibody

Description
Immunogen sequence: AESPKTPSSP LTPTFASSMC LLAPCYLTGD SVRDKCVEML SAALKADDDY KDYGVNCDKM ASEIEDH Highest antigen sequence identity to the following orthologs: Mouse - 84%, Rat - 85%.
Specifications
Specifications
| Antigen | TCEA3 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.6 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | TCEA3 |
| Gene Accession No. | O75764 |
| Gene Alias | hypothetical protein LOC402983; MGC137536 protein; rhabdomyosarcoma antigen MU-RMS-40.22; S-II; SII-K1; Tcea3; TFIIS; TFIIS.H; Tfiish; tissue-specific transcription factor S-II; transcription elongation factor A (SII), 3; transcription elongation factor A protein 3; transcription elongation factor A3; transcription elongation factor S-II protein 3; Transcription elongation factor TFIIS.h; zgc:77732 |
| Gene Symbols | TCEA3 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.