Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TCEA3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA569489
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA569489 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA569489 Supplier Invitrogen™ Supplier No. PA569489
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 100%; Dog: 93%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 79%.

Necessary for efficient RNA polymerase II transcription elongation past template-encoded arresting sites. The arresting sites in DNA have the property of trapping a certain fraction of elongating RNA polymerases that pass through, resulting in locked ternary complexes. Cleavage of the nascent transcript by S-II allows the resumption of elongation from the new 3'-terminus.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TCEA3
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene TCEA3
Gene Accession No. P23881
Gene Alias hypothetical protein LOC402983; MGC137536 protein; rhabdomyosarcoma antigen MU-RMS-40.22; S-II; SII-K1; Tcea3; TFIIS; TFIIS.H; Tfiish; tissue-specific transcription factor S-II; transcription elongation factor A (SII), 3; transcription elongation factor A protein 3; transcription elongation factor A3; transcription elongation factor S-II protein 3; Transcription elongation factor TFIIS.h; zgc:77732
Gene Symbols TCEA3
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of human TCEA3 (aa 244-293).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 21401
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.