Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TCEAL1 Monoclonal Antibody (OTI2G10)
GREENER_CHOICE

Catalog No. PIMA526910
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA526910 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA526910 Supplier Invitrogen™ Supplier No. MA526910
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TCEAL1 Monoclonal Antibody for Western Blot, IHC (P)

TCEAL1 (transcription elongation factor A (SII) protein 3), also known as p21, SIIR or pp21, is a member of the TFS-II family. Transcription elongation factors of the TFS-II family are responsible for releasing RNA polymerase II (Pol II) from transcriptional arrest. DNA arresting sites can result in locked ternary complexes if elongating RNA polymerases are trapped. Transcription elongation factors function to activate the intrinsic RNA cleavage activity of RNA polymerases. This allows the RNA polymerase to cleave the nascent transcript, thereby forming a new 3'-terminus to resume elongation. TCEAL1 is a 157 amino acid nuclear protein that is ubiquitously expressed. TCEAL1 may exert its effects via protein-protein interactions with other transcriptional regulators rather than via direct binding of DNA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TCEAL1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI2G10
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene TCEAL1
Gene Accession No. Q15170
Gene Alias 0610011M09Rik; BC011290; Nuclear phosphoprotein p21/SIIR; P21; pp21; SIIR; TCEAL1; TCEA-like protein 1; transcription elongation factor A (SII)-like 1; transcription elongation factor A like 1; transcription elongation factor A protein-like 1; Transcription elongation factor S-II protein-like 1; WEX9
Gene Symbols TCEAL1
Host Species Mouse
Immunogen Full length human recombinant protein of TCEAL1 produced in E.coli.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9338
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.