Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TCEAL2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA589056
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA589056 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA589056 Supplier Invitrogen™ Supplier No. PA589056
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MEKLFNENEG MPSNQGKIDN EEQPPHEGKP EVACILEDKK LENEGNTENT GKRVEEPLKD KEKPESAGKA KGEGKSERKG KSEMQGGSKT EGKPERGGRA EGEGEPDSER; Positive Samples: Mouse brain; Cellular Location: Nucleus.

TCEAL2 is a member of the transcription elongation factor A (SII)-like (TCEAL) gene family. Members of this family contain TFA domains and may function as nuclear phosphoproteins that modulate transcription in a promoter context-dependent manner.This gene encodes a member of the transcription elongation factor A (SII)-like (TCEAL) gene family. Members of this family contain TFA domains and may function as nuclear phosphoproteins that modulate transcription in a promoter context-dependent manner. Multiple family members are located on the X chromosome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TCEAL2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.68 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene Alias my048; MY0876G0; MY0876G05; TCEAL2; TCEA-like protein 2; transcription elongation factor A (SII)-like 2; transcription elongation factor A like 2; transcription elongation factor A protein-like 2; Transcription elongation factor S-II protein-like 2; WEX1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-110 of human TCEAL2 (NP_5251291).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.