Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TCEAL3 Antibody, PerCP, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298537PCP
Description
TCEAL3 Polyclonal antibody specifically detects TCEAL3 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
TCEAL3 | |
Polyclonal | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
MGC15737, TCEA-like protein 3, transcription elongation factor A (SII)-like 3, transcription elongation factor A protein-like 3, Transcription elongation factor S-II protein-like 3 | |
Produced in rabbits immunized with E. coli-derived Human TCEAL3 fragment. | |
0.1 mL | |
DNA replication Transcription Translation and Splicing | |
85012 | |
Store at 4C in the dark. | |
IgG |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
PerCP | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction