Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TCEAL5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561034
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561034 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561034 Supplier Invitrogen™ Supplier No. PA561034
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PKDSQEDLQE RHLSSEEMMR ECGDVSRAQE ELRK Highest antigen sequence identity to the following orthologs: Mouse - 82%, Rat - 79%.

TCEAL5, which is located on the X chromosome, encodes a protein which contains a BEX (brain expressed X-liked like family) domain. This domain is found in proteins encoded by the TCEAL elongation factor (transcription elongation factor A (SII)-like) gene family also located on the X chromosome. The coding region for this gene is located entirely in the terminal exon.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TCEAL5
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TCEAL5
Gene Accession No. Q5H9L2
Gene Alias 6430401A05Rik; LOC100059142; Tceal5; TCEA-like protein 5; transcription elongation factor A (SII)-like 5; transcription elongation factor A like 5; transcription elongation factor A protein-like 5; Transcription elongation factor S-II protein-like 5; WEX4
Gene Symbols TCEAL5
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TCEAL3. Recombinant protein control fragment (Product #RP-104390).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 340543
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.