Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TCEAL3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563637
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563637 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563637 Supplier Invitrogen™ Supplier No. PA563637
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EGEPGDEGQL EDEGSQEKQG RSEGE Highest antigen sequence identity to the following orthologs: Mouse - 56%, Rat - 56%.

TCEAL3 encodes a member of the transcription elongation factor A (SII)-like (TCEAL) gene family. Members of this family contain TFA domains and may function as nuclear phosphoproteins that modulate transcription in a promoter context-dependent manner. Multiple family members are located on the X chromosome. Alternative splicing results in multiple transcript variants encoding a single isoform.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TCEAL3
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TCEAL3
Gene Accession No. Q969E4
Gene Alias 1100001D19Rik; MSTP072; TCEAL3; TCEA-like protein 3; transcription elongation factor A (SII)-like 3; transcription elongation factor A like 3; transcription elongation factor A protein-like 3; Transcription elongation factor S-II protein-like 3; WEX8
Gene Symbols TCEAL3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TCEAL3. Recombinant protein control fragment (Product #RP-104524).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 85012
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.