Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TCEAL3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5112290
Description
TCEAL3 Polyclonal Antibody for Western Blot, IHC (P)
TCEAL3 encodes a member of the transcription elongation factor A (SII)-like (TCEAL) gene family. Members of this family contain TFA domains and may function as nuclear phosphoproteins that modulate transcription in a promoter context-dependent manner. Multiple family members are located on the X chromosome. Alternative splicing results in multiple transcript variants encoding a single isoform.
Specifications
TCEAL3 | |
Polyclonal | |
Unconjugated | |
TCEAL3 | |
1100001D19Rik; MSTP072; TCEAL3; TCEA-like protein 3; transcription elongation factor A (SII)-like 3; transcription elongation factor A like 3; transcription elongation factor A protein-like 3; Transcription elongation factor S-II protein-like 3; WEX8 | |
Rabbit | |
Antigen Affinity Chromatography, Protein A | |
RUO | |
85012 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 0.03% ProClin 300; pH 7 | |
Q969E4 | |
TCEAL3 | |
E. coli-derived Human TCEAL3 fragment. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction