Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TCEAL4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA567536
Description
TCEAL4 Polyclonal Antibody for Western Blot
May be involved in transcriptional regulation. Lu X.W., Zhongguo Yi Xue Ke Xue Yuan Xue Bao 23:27-31(2001). Xu X., Submitted (MAY-2000) to the EMBL/GenBank/DDBJ databases. Ota T., Nat. Genet. 36:40-45(2004).
Specifications
TCEAL4 | |
Polyclonal | |
Unconjugated | |
TCEAL4 | |
NPD017; TCEAL4; TCEA-like protein 4; transcription elongation factor A (SII)-like 4; transcription elongation factor A like 4; transcription elongation factor A protein-like 4; Transcription elongation factor S-II protein-like 4; WEX7 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
79921 | |
-20°C | |
Liquid |
Western Blot | |
0.6 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
Q96EI5 | |
TCEAL4 | |
Full length Fusion protein of human TCEAL4. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction