Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TCEAL5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5104185
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5104185 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5104185 Supplier Invitrogen™ Supplier No. PA5104185
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total TCEAL5.

TCEAL5, which is located on the X chromosome, encodes a protein which contains a BEX (brain expressed X-liked like family) domain. This domain is found in proteins encoded by the TCEAL elongation factor (transcription elongation factor A (SII)-like) gene family also located on the X chromosome. The coding region for this gene is located entirely in the terminal exon.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TCEAL5
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene TCEAL5
Gene Accession No. Q5H9L2, Q8CCT4
Gene Alias 6430401A05Rik; LOC100059142; Tceal5; TCEA-like protein 5; transcription elongation factor A (SII)-like 5; transcription elongation factor A like 5; transcription elongation factor A protein-like 5; Transcription elongation factor S-II protein-like 5; WEX4
Gene Symbols TCEAL5
Host Species Rabbit
Immunogen A synthesized peptide derived from human TCEAL5(Accession Q5H9L2), corresponding to amino acid residues S131-P181.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 331532, 340543, 680282
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.