Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TCEAL7 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA568350
Description
Predicted to react with Mouse samples.
Plays a role in the negative regulation of NF-kappa-B signaling at the basal level by modulating transcriptional activity of NF-kappa-B on its target gene promoters. Associates with cyclin D1 promoter containing Myc E-box sequence and transcriptionally represses cyclin D1 expression. Regulates telomerase reverse transcriptase expression and telomerase activity in both ALT (alternative lengthening of telomeres)and telomerase-positive cell lines.
Specifications
TCEAL7 | |
Polyclonal | |
Unconjugated | |
TCEAL7 | |
1110018P05Rik; MPMGp800C04260Q003; TCEAL7; TCEA-like protein 7; transcription elongation factor A (SII)-like 7; transcription elongation factor A like 7; transcription elongation factor A protein-like 7; Transcription elongation factor S-II protein-like 7; WEX5 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
100040972, 56849 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
A3KGA4, Q9BRU2 | |
TCEAL7 | |
Synthesized peptide derived from human TCEAL7. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction