Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TCEAL7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5104186
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5104186 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5104186 Supplier Invitrogen™ Supplier No. PA5104186
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total TCEAL7.

Plays a role in the negative regulation of NF-kappa-B signaling at the basal level by modulating transcriptional activity of NF-kappa-B on its target gene promoters. Associates with cyclin D1 promoter containing Myc E-box sequence and transcriptionally represses cyclin D1 expression. Regulates telomerase reverse transcriptase expression and telomerase activity in both ALT (alternative lengthening of telomeres)and telomerase-positive cell lines.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TCEAL7
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene TCEAL7
Gene Accession No. A3KGA4, Q9BRU2
Gene Alias 1110018P05Rik; MPMGp800C04260Q003; TCEAL7; TCEA-like protein 7; transcription elongation factor A (SII)-like 7; transcription elongation factor A like 7; transcription elongation factor A protein-like 7; Transcription elongation factor S-II protein-like 7; WEX5
Gene Symbols TCEAL7
Host Species Rabbit
Immunogen A synthesized peptide derived from human TCEAL7(Accession Q9BRU2), corresponding to amino acid residues R23-L73.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100040972, 56849
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.