Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TCEAL8 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA543549
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA543549 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA543549 Supplier Invitrogen™ Supplier No. PA543549
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: PQPSEEGVSQ EAEGNPRGGP NQPGQGFKED TPVRHLDPEE MIRGVDELER Sequence homology: Human: 100%.

TCEAL8 is a member of the transcription elongation factor A (SII)-like (TCEAL) gene family. Members of this family contain TFA domains and may function as nuclear phosphoproteins that modulate transcription in a promoter context-dependent manner. This gene encodes a member of the transcription elongation factor A (SII)-like (TCEAL) gene family. Members of this family contain TFA domains and may function as nuclear phosphoproteins that modulate transcription in a promoter context-dependent manner. Multiple family members are located on the X chromosome. Alternative splicing results in multiple transcript variants encoding a single isoform.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TCEAL8
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene TCEAL8
Gene Accession No. Q8IYN2
Gene Alias 3930402F23Rik; MGC45400; TCEAL8; TCEA-like protein 8; transcription elongation factor A (SII)-like 8; transcription elongation factor A like 8; transcription elongation factor A protein-like 8; Transcription elongation factor S-II protein-like 8; up-regulated in nephrectomized rat kidney #1; UR-NR#1; WEX3
Gene Symbols TCEAL8
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human TCEAL8.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 90843
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.